Learn More
CD14, Mouse, PE-Cy5.5, Clone: 61D3, Abnova™
Mouse monoclonal antibody raised against native CD14.
Brand: Abnova MAB6045.100Reactions
Description
CD14 is a surface protein preferentially expressed on monocytes/macrophages. It binds lipopolysaccharide binding protein and recently has been shown to bind apoptotic cells. Alternative splicing results in multiple transcript variants encoding the same isoform. [provided by RefSeq
Specifications
| CD14 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| Flow Cytometry (10 ul/106 cells) The optimal working dilution should be determined by the end user. | |
| CD14 | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| Specificity human CD14 | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| 61D3 | |
| Mouse monoclonal antibody raised against native CD14. | |
| In PBS (0.09% sodium azide) | |
| CD14 | |
| Native purified from human peripheral blood mononuclear cells. | |
| RUO | |
| 929 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.