missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD175, Mouse, Clone: BRIC111, Abnova™
Mouse monoclonal antibody raised against native CD175.
Brand: Abnova MAB5231.200ug
This item is not returnable.
View return policy
Specifications
| CD175 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 200 μg | |
| Primary | |
| Human | |
| Liquid |
| Agglutination, Immunohistochemistry (Frozen) | |
| BRIC111 | |
| Mouse monoclonal antibody raised against native CD175. | |
| In Tris-glycine, pH 7.4 (0.09% sodium azide) | |
| Native purified CD175 from human Tn red blood cells. | |
| RUO | |
| This antibody recognizes Tn antigen on glycophorin A and glycophorin B in human erythrocytes. Tn is a cryptantigen which was recently designated CD175 at the 7th Leucocyte Typing Workshop. Tn antigen is not expressed on normal haemopoietic cells but exposure of the Tn is associated with polyagglutination. | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction