missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19, Mouse, APC, Clone: SJ25-C1, Abnova™
Mouse monoclonal antibody raised against native CD19.
Brand: Abnova MAB5981.100Reactions
This item is not returnable.
View return policy
Specifications
| CD19 | |
| Monoclonal | |
| APC | |
| Flow Cytometry (10 ul/106 cells) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen), Immunoprecipitation | |
| SJ25-C1 | |
| Mouse monoclonal antibody raised against native CD19. | |
| In PBS (0.09% sodium azide) | |
| Native purified from NALM-1 and NALM-16 cells. | |
| RUO | |
| Specificity CD19 | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction