missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19, Mouse, FITC, Clone: MB19-1, Abnova™
Mouse monoclonal antibody raised against Cd19.
Brand: Abnova MAB5695.500ug
This item is not returnable.
View return policy
Specifications
| CD19 | |
| Monoclonal | |
| FITC | |
| Flow Cytometry (1 ug/106 cells) The optimal working dilution should be determined by the end user. | |
| Cd19 | |
| Cd19 | |
| 300.191., CD19+ mouse pre-B cell line | |
| RUO | |
| 12478 | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunoprecipitation | |
| MB19-1 | |
| Mouse monoclonal antibody raised against Cd19. | |
| In PBS (0.09% sodium azide) | |
| AW495831 | |
| Mouse | |
| 500 μg | |
| Primary | |
| CD19 B cell differentiation antigen, Mr 95 KDa. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgA κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction