missing translation for 'onlineSavingsMsg'
Learn More

CD2, Mouse, PE, Clone: MEM-65, Abnova™

Product Code. 16066356
Change view
Click to view available options
Quantity:
100 Reactions
Unit Size:
100 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16066356 100 Reactions 100 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16066356 Supplier Abnova Supplier No. MAB4558.100Reactions

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against native CD2.

Specifications

Antigen CD2
Applications Flow Cytometry
Classification Monoclonal
Clone MEM-65
Conjugate PE
Description Mouse monoclonal antibody raised against native CD2.
Dilution Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.2% BSA, 0.09% sodium azide)
Gene CD2
Gene Alias SRBC/T11
Gene Symbols CD2
Host Species Mouse
Immunogen Native purified CD2 from human peripheral T cells.
Quantity 100 Reactions
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 914
Test Specificity This antibody recognizes an unique epitope of CD2, a 50 KDa glycoprotein present on the human peripheral blood T-lymphocytes and NK cells; also expressed by all thymocytes.
Target Species Human
Content And Storage Store in the dark at 4°C. Do not freeze.
Avoid prolonged exposure to light.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.