missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD24, Mouse, PE, Clone: SN3, Abnova™
Mouse monoclonal antibody raised against CD24.
Brand: Abnova MAB4395.100Reactions
This item is not returnable.
View return policy
Specifications
| CD24 | |
| Monoclonal | |
| PE | |
| Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user. | |
| CD24 | |
| CD24 | |
| Human CD24. | |
| RUO | |
| 100133941 | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| SN3 | |
| Mouse monoclonal antibody raised against CD24. | |
| In PBS (0.2% BSA, 0.09% sodium azide) | |
| CD24A/FLJ22950/FLJ43543/MGC75043 | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| This antibody reacts with CD24, a 35-45 KDa heavily glycosylated cell surface antigen. CD24 is expressed by granulocytes, B lymphocytes and by some activated T cells and T cell malignancies. It is not expressed on human thymocytes. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction