missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD34, Mouse, PE, Clone: 4H11[APG], Abnova™
Mouse monoclonal antibody raised against native CD34.
Brand: Abnova MAB4413.100Reactions
This item is not returnable.
View return policy
Description
CD34 is a monomeric cell surface antigen with a molecular mass of approximately 110kDa that is selectively expressed on human hematopoietic progenitor cells.[supplied by OMIM
Specifications
| CD34 | |
| Monoclonal | |
| PE | |
| Flow Cytometry (20 ul in human blood cells 100 ul in whole blood or 106 cells in a suspension) The optimal working dilution should be determined by the end user. | |
| CD34 | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| This antibody reacts with Class III epitope on CD34 (Mucosialin), a 110-115 KDa monomeric transmembrane phosphoglycoprotein expressed on hematopoietic progenitors cells and on the most pluripotential stem cells; it is gradually lost on progenitor cells. | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Flow Cytometry, Western Blot | |
| 4H11[APG] | |
| Mouse monoclonal antibody raised against native CD34. | |
| In PBS (0.2% BSA, 0.09% sodium azide) | |
| CD34 | |
| Native purified CD34 from permanent human cell line derived from peripheral leucocytes of a patient suffering from chronic myeloid leukaemia. | |
| RUO | |
| 947 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction