Learn More
HPB-ALL, Mouse, Clone: MEM-258, Abnova™
Description
The protein encoded by this gene is a type I membrane protein and is a regulatory part of the complement system. The encoded protein has cofactor activity for inactivation of complement components C3b and C4b by serum factor I, which protects the host cell from damage by complement. In addition, the encoded protein can act as a receptor for the Edmonston strain of measles virus, human herpesvirus-6, and type IV pili of pathogenic Neisseria. Finally, the protein encoded by this gene may be involved in the fusion of the spermatozoa with the oocyte during fertilization. This gene is found in a cluster on chromosome 1q32 with other genes encoding structural components of the complement system. At least fourteen different transcript variants encoding fourteen different isoforms have been found for this gene. [provided by RefSeq
Specifications
Specifications
| Antigen | HPB-ALL |
| Applications | Flow Cytometry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | MEM-258 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against native CD46. |
| Dilution | The optimal working dilution should be determined by the end user. |
| Formulation | In TBS, pH 8.0 (0.09% sodium azide) |
| Gene | CD46 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.