Learn More
CD57 Mouse anti-Human, BB515, Clone: NK-1, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 565285
Description
The NK-1 monoclonal antibody specifically reacts with a 110 kDa carbohydrate antigen associated with myelin-associated glycoprotein expressed on 7-35% of normal peripheral blood lymphocytes including a subset of natural killer cells, a subset of CD8-positive peripheral blood T cells, and on some neural tissues. CD57 is not expressed on granulocytes, platelets, red blood cells or thymocytes. The function of CD57 is still unclear, however, its expression on T-cell subets occurs in late immune responses.
The antibody was conjugated to BD Horizon BB515 which was developed exclusively by BD Biosciences. With an excitation max of 490nm and an emission max of 515nm, BD Horizon BB515 can be excited by the 488nm laser and detected in a standard FITC set (e.g. 530/30-nm filter). This dye provides a much brighter alternative to FITC with less spillover into the PE detector.
Specifications
| CD57 | |
| Monoclonal | |
| Brilliant Blue 515 (BB515) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgM κ |
| Flow Cytometry | |
| NK-1 | |
| B3GAT1; Beta-1,3-glucuronyltransferase 1; HNK1; LEU7; NK-1; GLCATP; GLCUATP | |
| Mouse | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.