Learn More
CD79B, Mouse, APC, Clone: CB3-1, Abnova™
Mouse monoclonal antibody raised against native CD79B.
Brand: Abnova MAB6084.100Reactions
Description
The B lymphocyte antigen receptor is a multimeric complex that includes the antigen-specific component, surface immunoglobulin (Ig). Surface Ig noncovalently associates with two other proteins, Ig-alpha and Ig-beta, which are necessary for expression and function of the B-cell antigen receptor. This gene encodes the Ig-beta protein of the B-cell antigen component. Alternatively spliced transcript variants encoding different isoforms have been described. [provided by RefSeq
Specifications
| CD79B | |
| Monoclonal | |
| APC | |
| Flow Cytometry (10 ul/106 cells) The optimal working dilution should be determined by the end user. | |
| CD79B | |
| CD79B | |
| Native purified from Ramos cells. | |
| RUO | |
| 974 | |
| Human | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Frozen) | |
| CB3-1 | |
| Mouse monoclonal antibody raised against native CD79B. | |
| In PBS (0.09% sodium azide) | |
| B29/IGB | |
| Mouse | |
| 100 Reactions | |
| Primary | |
| Specificity human CD79beta/B29/Igbeta | |
| Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.