missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD8, Mouse, Clone: MEM-87, Abnova™
Mouse monoclonal antibody raised against native CD8.
Brand: Abnova MAB0936.100ug
This item is not returnable.
View return policy
Specifications
| CD8 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CD8/Leu2/MAL/p32 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody recognizes CD8, a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. CD8 is a disulfide-linked dimer and exists as a CD8 alpha; homodimer or CD8 alpha/beta heterodimer (each monomer approx. 32-34 KDa). | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Flow Cytometry, Immunoprecipitation | |
| MEM-87 | |
| Mouse monoclonal antibody raised against native CD8. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| CD8A | |
| Native purified CD8 from leukocytes of a patient suffering from a LGL-type leukemia. | |
| RUO | |
| 925/926 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction