Learn More
CD8A, Mouse, Clone: SPM548, Abnova™
Mouse monoclonal antibody raised against CD8A.
Brand: Abnova MAB12029.100ug
Description
The CD8 antigen is a cell surface glycoprotein found on most cytotoxic T lymphocytes that mediates efficient cell-cell interactions within the immune system. The CD8 antigen acts as a corepressor with the T-cell receptor on the T lymphocyte to recognize antigens displayed by an antigen presenting cell (APC) in the context of class I MHC molecules. The coreceptor functions as either a homodimer composed of two alpha chains, or as a heterodimer composed of one alpha and one beta chain. Both alpha and beta chains share significant homology to immunoglobulin variable light chains. This gene encodes the CD8 alpha chain isoforms. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| CD8A | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CD8A | |
| CD8A | |
| Recombinant protein corresponding to human CD8A. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| SPM548 | |
| Mouse monoclonal antibody raised against CD8A. | |
| In 10mM PBS (0.05% BSA, 0.05% sodium azide) | |
| CD8/Leu2/MAL/p32 | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 925 | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.