missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD9, Mouse, Clone: IVA50, Abnova™
Mouse monoclonal antibody raised against native CD9.
Brand: Abnova MAB0985.100ug
This item is not returnable.
View return policy
Specifications
| CD9 | |
| Monoclonal | |
| 1 mg/mL | |
| Mouse monoclonal antibody raised against native CD9. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| CD9 | |
| Native purified CD9 from bovine thrombocytes. | |
| RUO | |
| 280746 | |
| Store at 4°C. Do not freeze. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2 |
| Flow Cytometry, Immunoprecipitation, Western Blot | |
| IVA50 | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CD9 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Bovine | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction