Learn More
CDC2, Mouse, Clone: POH-1, Abnova™
Mouse monoclonal antibody raised against recombinant CDC2.
Brand: Abnova MAB1921.100ug
Description
The protein encoded by this gene is a member of the Ser/Thr protein kinase family. This protein is a catalytic subunit of the highly conserved protein kinase complex known as M-phase promoting factor (MPF), which is essential for G1/S and G2/M phase transitions of eukaryotic cell cycle. Mitotic cyclins stably associate with this protein and function as regulatory subunits. The kinase activity of this protein is controlled by cyclin accumulation and destruction through the cell cycle. The phosphorylation and dephosphorylation of this protein also play important regulatory roles in cell cycle control. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Specifications
| CDC2 | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:500-1:1000) Immunohistochemistry (1:50) The optimal working dilution should be determined by the end user. | |
| CDC2 | |
| CDC2 | |
| Recombinant protein corresponding to human CDC2. | |
| RUO | |
| 983 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG2a κ |
| ELISA, Immunoprecipitation, Western Blot | |
| POH-1 | |
| Mouse monoclonal antibody raised against recombinant CDC2. | |
| In 20mM KH2PO4, 500mM NaCl, pH 7.2 (0.01% sodium azide) | |
| CDC28A/CDK1/DKFZp686L20222/MGC111195 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This protein A purified mouse monoclonal antibody reacts specifically with p34 cdc2 in human tissues and cell lines. This antibody is not cross reactive with other cyclin dependent kinases. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.