Learn More
CDC25C, Mouse, Clone: 3B11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CDC25C.
Brand: Abnova H00000995-M01A.200uL
Description
This gene is highly conserved during evolution and it plays a key role in the regulation of cell division. The encoded protein is a tyrosine phosphatase and belongs to the Cdc25 phosphatase family. It directs dephosphorylation of cyclin B-bound CDC2 and triggers entry into mitosis. It is also thought to suppress p53-induced growth arrest. Multiple alternatively spliced transcript variants of this gene have been described, however, the full-length nature of many of them is not known. [provided by RefSeq
Sequence: FRSNQRKMLNLLLERDTSFTVCPDVPRTPVGKFLGDSANLSILSGGTPKCCLDLSNLSSGEITATQLTTSADLDETGHLDSSGLQEVHLAGMNHDQHLMKCSPAQLLCSTSpecifications
| CDC25C | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC019089 | |
| CDC25C | |
| CDC25C (AAH19089,21 a.a. ∽ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 995 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 3B11 | |
| Mouse monoclonal antibody raised against a partial recombinant CDC25C. | |
| CDC25C | |
| CDC25 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.