missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CDC42BPG, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CDC42BPG.
Brand: Abnova H00055561-A01.50uL
This item is not returnable.
View return policy
Description
Sequence: RREMLKDPFVRSKLISPPTNFNHLVHVGPANGRPGARDKSPAPEEKGRVARGSGPQRPHSFSEALRRPASMGSEGLGGDADPMKRKPWTSLSSESVSCPQGSLSPASpecifications
| CDC42BPG | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CDC42BPG. | |
| CDC42BPG | |
| DMPK2/HSMDPKIN/KAPPA-200/MRCKgamma | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_017525 | |
| CDC42BPG | |
| CDC42BPG (NP_059995, 1423 a.a. to 1528 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 55561 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction