missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdh1, Mouse, Clone: M168, Abnova™
Mouse monoclonal antibody raised against partial recombinant Cdh1.
Brand: Abnova MAB1388.100uL
This item is not returnable.
View return policy
Specifications
| Cdh1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:2000) Immunocytochemistry (1:250) Immunoprecipitation (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| Cdh1 | |
| Cdh1 | |
| Recombinant protein corresponding to C-terminus of mouse Cdh1. | |
| RUO | |
| 12550 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
| M168 | |
| Mouse monoclonal antibody raised against partial recombinant Cdh1. | |
| In PBS (50% glycerol, 1mg/mL BSA, 0.05% sodium azide) | |
| AA960649/Ecad/L-CAM/MGC107495/UVO/Um | |
| Mouse | |
| 100 μL | |
| Primary | |
| This sequence is highly conserved in human and rat E-cadherin. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction