Learn More
CDKN1C, Mouse, Clone: KP10, Abnova™
Mouse monoclonal antibody raised against CDKN1C.
Brand: Abnova MAB10749.1mL
Description
The protein encoded by this gene is a tight-binding, strong inhibitor of several G1 cyclin/Cdk complexes and a negative regulator of cell proliferation. Mutations in this gene are implicated in sporadic cancers and Beckwith-Wiedemann syndorome, suggesting that this gene is a tumor suppressor candidate. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq
Specifications
| CDKN1C | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CDKN1C | |
| CDKN1C | |
| CDKN1C. | |
| RUO | |
| 1028 | |
| Liquid |
| Immunohistochemistry | |
| KP10 | |
| Mouse monoclonal antibody raised against CDKN1C. | |
| In PBS, pH 7.4 (protein base, 0.09% sodium azide) | |
| BWCR/BWS/KIP2/WBS/p57 | |
| Mouse | |
| 1 mL | |
| Primary | |
| Store at 4°C is stable for 3 years. | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.