missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CEACAM5, Mouse, Clone: C66/261, Abnova™
Mouse monoclonal antibody raised against CEACAM5.
Brand: Abnova MAB11368.100ug
This item is not returnable.
View return policy
Specifications
| CEACAM5 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1 ug/million cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (0.5-1 ug/mL for 30 min at RT) Immunoprecipitation (1-2 ug/500 ug protein) Western Blot (0.5-1 ug/mL) The optimal working dilution should be deter | |
| CD66e/CEA/DKFZp781M2392 | |
| Mouse | |
| Protein A/G affinity chromatography | |
| RUO | |
| 1048/634 | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
| C66/261 | |
| Mouse monoclonal antibody raised against CEACAM5. | |
| In PBS, pH 7.4 (0.05% BSA, 0.05% sodium azide) | |
| CEACAM5 | |
| Human colon carcinoma extract. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction