Learn More
CGA, Mouse, Clone: HCGa/53, Abnova™
Mouse monoclonal antibody raised against recombinant human CGA.
Brand: Abnova MAB12127.100ug
Description
The four human glycoprotein hormones chorionic gonadotropin (CG), luteinizing hormone (LH), follicle stimulating hormone (FSH), and thyroid stimulating hormone (TSH) are dimers consisting of alpha and beta subunits that are associated noncovalently. The alpha subunits of these hormones are identical, however, their beta chains are unique and confer biological specificity. The protein encoded by this gene is the alpha subunit and belongs to the glycoprotein hormones alpha chain family. [provided by RefSeq
Specifications
| CGA | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (0.5-1 ug/mL) Western blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CGA | |
| CGA | |
| Recombinant protein corresponding to human CGA. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| Immunohistochemistry (PFA fixed), Western Blot | |
| HCGa/53 | |
| Mouse monoclonal antibody raised against recombinant human CGA. | |
| In PBS (0.05% BSA, 0.05% sodium azide) | |
| CG-ALPHA/FSHA/GPHA1/GPHa/HCG/LHA/TSHA | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 1081 | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.