Learn More
CGB Mouse anti-Human, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native CGB.
Brand: Abnova MAB1406.1mg
Description
This gene is a member of the glycoprotein hormone beta chain family and encodes the beta 3 subunit of chorionic gonadotropin (CG). Glycoprotein hormones are heterodimers consisting of a common alpha subunit and an unique beta subunit which confers biological specificity. CG is produced by the trophoblastic cells of the placenta and stimulates the ovaries to synthesize the steroids that are essential for the maintenance of pregnancy. The beta subunit of CG is encoded by 6 genes which are arranged in tandem and inverted pairs on chromosome 19q13.3 and contiguous with the luteinizing hormone beta subunit gene. [provided by RefSeq
Specifications
| CGB | |
| Monoclonal | |
| Mouse monoclonal antibody raised against native CGB. | |
| In 15mM KPO4 buffer, 0.85% NaCl, pH 7.4 (0.09% sodium azide) | |
| CGB3/hCGB | |
| Mouse | |
| 1 mg | |
| Primary | |
| This antibody reacts with CGA and CGB. | |
| Liquid |
| ELISA | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| CGB | |
| CGB | |
| Native purified human CGB. | |
| RUO | |
| 1082 | |
| Store at 4°C. | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.