Learn More
CHGA, Mouse, Clone: SPM585, Abnova™
Mouse monoclonal antibody raised against CHGA.
Brand: Abnova MAB12042.100ug
Description
The protein encoded by this gene is a member of the chromogranin/secretogranin family of neuroendocrine secretory proteins. It is found in secretory vesicles of neurons and endocrine cells. This gene product is a precursor to three biologically active peptides; vasostatin, pancreastatin, and parastatin. These peptides act as autocrine or paracrine negative modulators of the neuroendocrine system. Other peptides, including chromostatin, beta-granin, WE-14 and GE-25, are also derived from the full-length protein. However, biological activities for these molecules have not been shown. [provided by RefSeq
Specifications
| CHGA | |
| Monoclonal | |
| Unconjugated | |
| Immunohistochemistry (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CHGA | |
| CHGA | |
| Human pheochromocytoma. | |
| 100 μg | |
| Primary | |
| Does not react with mouse and rat. | |
| Store at 4°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| Immunohistochemistry (PFA fixed), Western Blot | |
| SPM585 | |
| Mouse monoclonal antibody raised against CHGA. | |
| In 10mM PBS (0.05% BSA, 0.05% sodium azide) | |
| CGA | |
| Mouse | |
| Protein A/G purification | |
| RUO | |
| 1113 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.