Learn More
CLOCK, Mouse, Clone: 8F7, Abnova™
Mouse monoclonal antibody raised against partial recombinant CLOCK.
Brand: Abnova MAB10570.100uL
Additional Details : Weight : 0.02000kg
Description
This gene encodes a protein that belongs to the basic helix-loop-helix (bHLH) family of transcription factors. Polymorphisms within the encoded protein have been associated with circadian rhythm sleep disorders. A similar protein in mice is a circadian regulator that acts as a transcription factor and forms a heterodimer with aryl hydrocarbon receptor nuclear translocator-like to activate transcription of mouse period 1. [provided by RefSeq
Specifications
CLOCK | |
Monoclonal | |
Unconjugated | |
ELISA (1:10000) Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
CLOCK | |
CLOCK | |
Recombinant protein corresponding to human CLOCK. | |
RUO | |
9575 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG1 |
ELISA, Immunofluorescence, Western Blot | |
8F7 | |
Mouse monoclonal antibody raised against partial recombinant CLOCK. | |
In ascites (0.03% sodium azide) | |
KAT13D/KIAA0334/bHLHe8 | |
Mouse | |
100 μL | |
Primary | |
Human | |
Liquid |