missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CLU, Mouse, Clone: 1A11, Abnova™
Mouse monoclonal antibody raised against partial recombinant CLU.
Brand: Abnova MAB1098.100uL
This item is not returnable.
View return policy
Specifications
| CLU | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user. | |
| CLU | |
| AAG4/APOJ/CLI/KUB1/MGC24903/SGP-2/SGP2/SP-40/TRPM-2/TRPM2 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Immunohistochemistry, Western Blot | |
| 1A11 | |
| Mouse monoclonal antibody raised against partial recombinant CLU. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| NM_203339 | |
| CLU | |
| Recombinant protein corresponding to amino acids 1-333 of human CLU. | |
| RUO | |
| 1191 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction