Learn More
COG4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant COG4.
Brand: Abnova H00025839-A01.50uL
Description
Multiprotein complexes are key determinants of Golgi apparatus structure and its capacity for intracellular transport and glycoprotein modification. Several complexes have been identified, including the Golgi transport complex (GTC), the LDLC complex, which is involved in glycosylation reactions, and the SEC34 complex, which is involved in vesicular transport. These 3 complexes are identical and have been termed the conserved oligomeric Golgi (COG) complex, which includes COG4 (Ungar et al., 2002 [PubMed 11980916]).[supplied by OMIM
Sequence: ELEKVVLKSTFNRLGGLQFDKELRSLIAYLTTVTTWTIRDKFARLSQMATILNLERVTEILDYWGPNSGPLTWRLTPAEVRQVLALRIDFRSEDIKRLRLSpecifications
| COG4 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant COG4. | |
| COG4 | |
| COD1/DKFZp586E1519 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_015386 | |
| COG4 | |
| COG4 (NP_056201, 686 a.a. to 785 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 25839 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only