Learn More
COL21A1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant COL21A1.
Brand: Abnova H00081578-A01.50uL
Description
This gene encodes the alpha chain of type XXI collagen, a member of the FACIT collagen family (fibril-associated collagens with interrupted helices). Type XXI collagen is localized to tissues containing type I collagen so, like other members of this collagen family, it may serve to maintain the integrity of the extracellular matrix. An alternatively spliced transcript variant has been described, but its full-length nature has yet to be determined. [provided by RefSeq
Sequence: IPVAARDERGFDILLGLDVNKKVKKRIQLSPKKIKGYEVTSKVDLSELTSNVFPEGLPPSYVFVSTQRFKVKKIWDLWRILTIDGRPQIAVTLNGVDKILSpecifications
| COL21A1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant COL21A1. | |
| COL21A1 | |
| COLA1L/DKFZp564B052/FLJ39125/FLJ44623/FP633/MGC26619/dJ682J15.1/dJ708F5.1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_030820 | |
| COL21A1 | |
| COL21A1 (NP_110447, 221 a.a. to 320 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 81578 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only