Learn More
CPNE7, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CPNE7.
Brand: Abnova H00027132-A01.50uL
Description
This gene encodes a member of the copine family, which is composed of calcium-dependent membrane-binding proteins. The gene product contains two N-terminal C2 domains and one von Willebrand factor A domain. The encoded protein may be involved in membrane trafficking. Two alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: ASRLPMSIIIVGVGNADFTDMQVLDGDDGVLRSPRGEPALRDIVQFVPFRELKNASPAALAKCVLAEVPKQVVEYYSHRGLPPRSLGVPAGEASPGCTPSpecifications
| CPNE7 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CPNE7. | |
| CPNE7 | |
| MGC34192 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_153636 | |
| CPNE7 | |
| CPNE7 (NP_705900, 460 a.a. to 558 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 27132 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only