missing translation for 'onlineSavingsMsg'
Learn More

CRP, Mouse, Clone: N1G1, Abnova™

Product Code. 16241570
Change view
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16241570 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16241570 Supplier Abnova Supplier No. MAB6379.500ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against native CRP.

Specifications

Antigen CRP
Applications ELISA, Western Blot
Classification Monoclonal
Clone N1G1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native CRP.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS, pH 7.2
Gene CRP
Gene Alias MGC149895/MGC88244/PTX1
Gene Symbols CRP
Host Species Mouse
Immunogen Native purified human pleural fluid CRP.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1401
Test Specificity This antibody react with human CRP.
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.