missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CSAD, Mouse, Clone: 2C11, Abnova™
Mouse monoclonal antibody raised against a partial recombinant CSAD.
Brand: Abnova H00051380-M02A.200uL
This item is not returnable.
View return policy
Description
Sequence: MADSEALPSLAGDPVAVEALLRAVFGVVVDEAIQKGTSVSQKVCEWKEPEELKQLLDLELRSQGESQKQILERCRAVIRYSVKTGHPRFFNQLFSGLDSpecifications
| CSAD | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_015989 | |
| CSAD | |
| CSAD (NP_057073,1 a.a. ∽ 98 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 51380 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 2C11 | |
| Mouse monoclonal antibody raised against a partial recombinant CSAD. | |
| CSAD | |
| CSD/FLJ44987/FLJ45500/MGC119354/MGC119355/MGC119357/PCAP | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction