Learn More
CST6, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant CST6.
Brand: Abnova H00001474-A01.50uL
Description
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions, where they appear to provide protective functions. This gene encodes a cystatin from the type 2 family, which is down-regulated in metastatic breast tumor cells as compared to primary tumor cells. Loss of expression is likely associated with the progression of a primary tumor to a metastatic phenotype. [provided by RefSeq
Sequence: MGSNSIYYFRDTHIIKAQSQLVAGIKYFLTMEMGSTDCRKTRVTGDHVDLTTCPLAAGAQQEKLRCDFEVLVVPWQNSSQLLKHNCVQMSpecifications
| CST6 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant CST6. | |
| CST6 | |
| CST6 | |
| CST6 (NP_001314, 61 a.a. to 149 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 1474 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001323 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only