Learn More
DCAMKL1, Mouse, Clone: 6F9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DCAMKL1.
Brand: Abnova H00009201-M02A.200uL
Description
DCAMKL1 is a microtubule-associated protein that phosphorylates itself and myelin basic protein (MBP; MIM 159430). DCAMKL1 has microtubule polymerizing activity that is independent of its protein kinase activity (Lin et al., 2000 [PubMed 11124993]).[supplied by OMIM
Sequence: QVLEHPWVNDDGLPENEHQLSVAGKIKKHFNTGPKPNSTAAGVSVIALDHGFTIKRSGSLDYYQQPGMYWIRPPLLIRRGRFSDEDATRMSpecifications
| DCAMKL1 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_004734 | |
| DCLK1 | |
| DCAMKL1 (NP_004725,640 a.a. ∽ 729 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9201 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 6F9 | |
| Mouse monoclonal antibody raised against a partial recombinant DCAMKL1. | |
| DCLK1 | |
| DCAMKL1/DCDC3A/DCLK/KIAA0369 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.