Learn More
DES, Mouse, Clone: 10H7D2, Abnova™
Mouse monoclonal antibody raised against partial recombinant DES.
Brand: Abnova MAB8300.100ug
Description
This gene encodes a muscle-specific class III intermediate filament. Homopolymers of this protein form a stable intracytoplasmic filamentous network connecting myofibrils to each other and to the plasma membrane. Mutations in this gene are associated with desmin-related myopathy, a familial cardiac and skeletal myopathy (CSM), and with distal myopathies. [provided by RefSeq
Specifications
| DES | |
| Monoclonal | |
| Unconjugated | |
| Western Blot (1:1000-1:2000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:1000-1:2000) Immunohistochemistry (Frozen sections) (1:1000-1:2000) ELISA (1:10000) The optimal working dilution should be determined by the e | |
| DES | |
| DES | |
| Recombinant protein corresponding to truncated human DES. | |
| RUO | |
| 1674 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot | |
| 10H7D2 | |
| Mouse monoclonal antibody raised against partial recombinant DES. | |
| In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide) | |
| CMD1I/CSM1/CSM2/FLJ12025/FLJ39719/FLJ41013/FLJ41793 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.