Learn More
DIAPH1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant DIAPH1.
Brand: Abnova H00001729-A01.50uL
Description
This gene is a homolog of the Drosophila diaphanous gene, and has been linked to autosomal dominant, fully penetrant, nonsyndromic sensorineural progressive low-frequency hearing loss. Actin polymerization involves proteins known to interact with diaphanous protein in Drosophila and mouse. It has therefore been speculated that this gene may have a role in the regulation of actin polymerization in hair cells of the inner ear. Alternatively spliced transcript variants encoding distinct isoforms have been found for this gene. [provided by RefSeq
Sequence: QFSEQVENIKPEIVSVTAACEELRKSESFSNLLEITLLVGNYMNAGSRNAGAFGFNISFLCKLRDTKSTDQKMTLLHFLAELCENDYPDVLKFPDELAHVEKASSpecifications
| DIAPH1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant DIAPH1. | |
| DIAPH1 | |
| DFNA1/DIA1/DRF1/FLJ25265/LFHL1/hDIA1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_005219 | |
| DIAPH1 | |
| DIAPH1 (NP_005210, 921 a.a. to 1024 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 1729 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only