Learn More
DNAJC7, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant DNAJC7.
Brand: Abnova H00007266-A01.50uL
Description
DNAJC7 belongs to the evolutionarily conserved DNAJ/HSP40 family of proteins, which regulate molecular chaperone activity by stimulating ATPase activity. DNAJ proteins may have up to 3 distinct domains: a conserved 70-amino acid J domain, usually at the N terminus; a glycine/phenylalanine (G/F)-rich region; and a cysteine-rich domain containing 4 motifs resembling a zinc finger domain (Ohtsuka and Hata, 2000 [PubMed 11147971]).[supplied by OMIM
Sequence: MAATEPELLDDQEAKREAETFKEQGNAYYAKKDYNEAYNYYTKAIDMCPKNASYYGNRAATLMMLGRFREALGDAQQSVRLDDSFVRGHLREGKCHLSLGNAMAACRSFQRALELDHKNAQAQQEFKNANAVMEYEKIAETDFEKRDFRKVVFCMDRALEFAPACHRFKILKAECLAMLGRYPEAQSVASDILRMDSTNADALYVRGLCLYYEDCIEKAVQFFVQALRMAPDHEKACIACRNAKALKAKKEDGNKAFKEGNYKLAYELYTEALGIDPNNIKTNAKLYCNRGTVNSKLRKLDDAIEDCTNAVKLDDTYIKAYLRRAQCYMDTEQYEEAVRDYEKVYQTEKTKEHKQLLKNAQLELKKSKRKDYYKILGVDKNASEDEIKKAYRKRALMHHPDRHSGASAEVQKEEEKKFKEVGEAFTILSDPKKKTRYDSGQDLDEEGMNMGDFDPNNIFKAFFGGPGGFSFEASGPGNFFFQFGSpecifications
| DNAJC7 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length recombinant DNAJC7. | |
| DNAJC7 | |
| DANJC7/DJ11/TPR2/TTC2 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| BC033772 | |
| DNAJC7 | |
| DNAJC7 (AAH33772, 1 a.a. to 484 a.a) full-length recombinant protein with GST tag. | |
| RUO | |
| 7266 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only