missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNP, Mouse, Clone: B489M, Abnova™
Mouse monoclonal antibody raised against DNP.
Brand: Abnova MAB6684.100ug
This item is not returnable.
View return policy
Specifications
| DNP | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 κ |
| ELISA | |
| B489M | |
| Mouse monoclonal antibody raised against DNP. | |
| In PBS (0.09% sodium azide) | |
| Did not use an immunogen; screened against DNP-BSA | |
| RUO | |
| Dinitrophenol (DNP)-conjugated proteins and peptides. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction