Learn More
EIF2AK4, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EIF2AK4.
Brand: Abnova H00440275-A01.50uL
Description
EIF2AK4 belongs to a family of kinases that phosphorylate the alpha subunit of eukaryotic translation initiation factor-2 (EIF2S1; MIM 603907) to downregulate protein synthesis in response to varied cellular stresses (Berlanga et al., 1999 [PubMed 10504407]).[supplied by OMIM
Sequence: VVSVGQMSMSRAINLTQKLWTAGITAEIMYDWSQSQEELQEYCRHHEITYVALVSDKEGSHVKVKSFEKERQTEKRVLETELVDHVLQKLRTKVTDERNGSpecifications
| EIF2AK4 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant EIF2AK4. | |
| EIF2AK4 | |
| GCN2/KIAA1338 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001013703 | |
| EIF2AK4 | |
| EIF2AK4 (NP_001013725, 1401 a.a. to 1500 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 440275 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only