missing translation for 'onlineSavingsMsg'
Learn More

EPHA1, Mouse, Clone: 5D2D1, Abnova™

Product Code. 16191070
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16191070 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16191070 Supplier Abnova Supplier No. MAB8303.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse monoclonal antibody raised against partial recombinant EPHA1.

This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. This gene is expressed in some human cancer cell lines and has been implicated in carcinogenesis. [provided by RefSeq

Specifications

Antigen EPHA1
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 5D2D1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant EPHA1.
Dilution Western Blot (1:200-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200-1:1000) Immunohistochemistry (Frozen sections) (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene EPHA1
Gene Alias EPH/EPHT/EPHT1/MGC163163
Gene Symbols EPHA1
Host Species Mouse
Immunogen Recombinant protein corresponding to truncated human EPHA1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2041
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.