Learn More
ERBB2IP, Mouse, Clone: 10G1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ERBB2IP.
Brand: Abnova H00055914-M01A.200uL
Description
This gene is a member of the leucine-rich repeat and PDZ domain (LAP) family. The encoded protein contains 17 leucine-rich repeats and one PDZ domain. It binds to the unphosphorylated form of the ERBB2 protein and regulates ERBB2 function and localization. It has also been shown to affect the Ras signaling pathway by disrupting Ras-Raf interaction. Alternate transcriptional splice variants encoding different isoforms have been found for this gene, but only two of them have been characterized to date. [provided by RefSeq
Sequence: QGHELAKQEIRVRVEKDPELGFSISGGVGGRGNPFRPDDDGIFVTRVQPEGPASKLLQPGDKIIQANGYSFINIEHGQAVSLLKTFQNTVELIIVREVSSSpecifications
| ERBB2IP | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| NM_018695 | |
| ERBB2IP | |
| ERBB2IP (NP_061165,1272 a.a. ∽ 1371 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 55914 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 10G1 | |
| Mouse monoclonal antibody raised against a partial recombinant ERBB2IP. | |
| ERBB2IP | |
| ERBIN/LAP2 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.