Learn More
ERCC6, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ERCC6.
Brand: Abnova H00002074-A01.50uL
Description
This gene encodes a DNA-binding protein that is important in transcription-coupled excision repair. The protein has ATP-stimulated ATPase activity; there are contradictory publications reporting presence or absence of helicase activity. The protein appears to interact with several transcription and excision repair proteins, and may promote complex formation at repair sites. Mutations in this gene result in Cockayne syndrome type B. [provided by RefSeq
Sequence: RARNHLILPERLESESGHLQEASALLPTTEHDDLLVEMRNFIAFQAHTDGQASTREILQEFESKLSASQSCVFRELLRNLCTFHRTSGGEGIWKLKPEYCSpecifications
| ERCC6 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant ERCC6. | |
| ERCC6 | |
| ARMD5/CKN2/COFS/COFS1/CSB/RAD26 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_000124 | |
| ERCC6 | |
| ERCC6 (NP_000115, 1394 a.a. to 1493 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 2074 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only