Learn More
EREG, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant EREG.
Brand: Abnova H00002069-A01.50uL
Description
Epiregulin is a member of the epidermal growth factor family. Epiregulin can function as a ligand of EGFR (epidermal growth factor receptor), as well as a ligand of most members of the ERBB (v-erb-b2 oncogene homolog) family of tyrosine-kinase receptors. [provided by RefSeq
Sequence: STTVIPSCIPGESSDNCTALVQTEDNPRVAQVSITKCSSDMNGYCLHGQCIYLVDMSQNYCRCEVGYTGVRCEHFFLTVHQPLSKESpecifications
| EREG | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant EREG. | |
| EREG | |
| ER | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001432 | |
| EREG | |
| EREG (NP_001423, 32 a.a. to 117 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 2069 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
For Research Use Only