Learn More
ETS1, Mouse, Clone: 10D2, Abnova™
Mouse monoclonal antibody raised against partial recombinant ETS1.
Brand: Abnova MAB10640.100uL
Description
ETS transcriptions factors, such as ETS1, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes (Dwyer et al., 2007 [PubMed 17986575]).[supplied by OMIM
Specifications
| ETS1 | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry (1:200-1:1000) The optimal working dilution should be determined by the end user. | |
| ETS1 | |
| ETS1 | |
| Recombinant protein corresponding to human ETS1. | |
| RUO | |
| 2113 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| ELISA, Immunohistochemistry, Western Blot | |
| 10D2 | |
| Mouse monoclonal antibody raised against partial recombinant ETS1. | |
| In ascites (0.03% sodium azide) | |
| ETS-1/EWSR2/FLJ10768 | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.