Learn More
F8 Mouse anti-Human, Monoclonal Antibody, Abnova™
Mouse monoclonal antibody raised against native F8.
Brand:  Abnova MAB2173.100ug
Product Code. 16075436
  • € 388.00 / 100µg
Description
This gene encodes coagulation factor VIII, which participates in the intrinsic pathway of blood coagulation; factor VIII is a cofactor for factor IXa which, in the presence of Ca+2 and phospholipids, converts factor X to the activated form Xa. This gene produces two alternatively spliced transcripts. Transcript variant 1 encodes a large glycoprotein, isoform a, which circulates in plasma and associates with von Willebrand factor in a noncovalent complex. This protein undergoes multiple cleavage events. Transcript variant 2 encodes a putative small protein, isoform b, which consists primarily of the phospholipid binding domain of factor VIIIc. This binding domain is essential for coagulant activity. Defects in this gene results in hemophilia A, a common recessive X-linked coagulation disorder. [provided by RefSeq]
Specifications
Specifications
F8
Monoclonal
Mouse monoclonal antibody raised against native F8.
In PBS, pH 7.4
AHF/DXS1253E/F8B/F8C/FVIII/HEMA
Mouse
100 μg
Primary
Recognize full-length human Factor VIII.
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG1
ELISA
Unconjugated
ELISA (1:32000) The optimal working dilution should be determined by the end user.
F8
F8
Native purified human F8.
RUO
2157