Learn More
FABP3, Mouse, Clone: 10E1, Abnova™
Mouse monoclonal antibody raised against native FABP3.
Brand: Abnova MAB0459.200ug
Description
The intracellular fatty acid-binding proteins (FABPs) belongs to a multigene family. FABPs are divided into at least three distinct types, namely the hepatic-, intestinal- and cardiac-type. They form 14-15kDa proteins and are thought to participate in the uptake, intracellular metabolism and/or transport of long-chain fatty acids. They may also be responsible in the modulation of cell growth and proliferation. Fatty acid-binding protein 3 gene contains four exons and its function is to arrest growth of mammary epithelial cells. This gene is a candidate tumor suppressor gene for human breast cancer. [provided by RefSeq
Specifications
| FABP3 | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| FABP3 | |
| FABP3 | |
| Native purified human FABP3. | |
| RUO | |
| 2170 | |
| Store at 4°C. | |
| IgG1 |
| ELISA | |
| 10E1 | |
| Mouse monoclonal antibody raised against native FABP3. | |
| In PBS, pH 7.4 (0.09% sodium azide) | |
| FABP11/H-FABP/MDGI/O-FABP | |
| Mouse | |
| 200 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.