missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHL1 (B01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human FHL1 protein.
Brand: Abnova H00002273-B01P.50ug
This item is not returnable.
View return policy
Description
LIM proteins, named for 'LIN11, ISL1, and MEC3,' are defined by the possession of a highly conserved double zinc finger motif called the LIM domain.[supplied by OMIM
Sequence: MAEKFDCHYCRDPLQGKKYVQKDGHHCCLKCFDKFCANTCVECRKPIGADSKEVHYKNRFWHDTCFRCAKCLHPLANETFVAKDNKILCNKCTTREDSPKCKGCFKAIVAGDQNVEYKGTVWHKDCFTCSNCKQVIGTGSFFPKGEDFYCVTCHETKFAKHCVKCNKAITSGGITYQDQPWHADCFVCVTCSKKLAGQRFTAVEDQYYCVDCYKNFVAKKCAGCKNPITGFGKGSSVVAYEGQSWHDYCFHCKKCSVNLANKRFVFHQEQVYCPDCAKKLSpecifications
| FHL1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human FHL1 protein. | |
| FHL1 | |
| FHL1B/FLH1A/KYO-T/MGC111107/SLIM1/XMPMA/bA535K18.1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2273 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_001449.3 | |
| FHL1 | |
| FHL1 (NP_001440.2,1 a.a. ∽ 280 a.a) full-length human protein. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction