missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FHL1 (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant FHL1.
Brand: Abnova H00002273-A01.50uL
This item is not returnable.
View return policy
Description
LIM proteins, named for 'LIN11, ISL1, and MEC3,' are defined by the possession of a highly conserved double zinc finger motif called the LIM domain.[supplied by OMIM
Sequence: DGHHCCLKCFDKFCANTCVECRKPIGADSKEVHYKNRFWHDTCFRCAKCLHPLANETFVAKDNKILCNKCTTREDSPKCKGCFKAIVAGDQNVEYKGTSpecifications
| FHL1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant FHL1. | |
| FHL1 | |
| FHL1B/FLH1A/KYO-T/MGC111107/SLIM1/XMPMA/bA535K18.1 | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_001449 | |
| FHL1 | |
| FHL1 (NP_001440, 23 a.a. to 120 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 2273 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction