Learn More
FOXQ1, Mouse, Clone: 3C1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FOXQ1.
Brand: Abnova H00094234-M25C.200uL
Description
FOXQ1 is a member of the FOX gene family, which is characterized by a conserved 110-amino acid DNA-binding motif called the forkhead or winged helix domain. FOX genes are involved in embryonic development, cell cycle regulation, tissue-specific gene expression, cell signaling, and tumorigenesis (Bieller et al., 2001 [PubMed 11747606]).[supplied by OMIM
Sequence: RDSAGGRLTLAEINEYLMGKFPFFRGSYTGWRNSVRHNLSLNDCFVKVLRDPSRPWGKDNYWMLNPNSEYTFADGVFRRRRKRLSHRAPVPSpecifications
| FOXQ1 | |
| Monoclonal | |
| Unconjugated | |
| tissue culture supernatant with no preservative | |
| NM_033260.3 | |
| FOXQ1 | |
| FOXQ1 (NP_150285.3,133 a.a. ∽ 223 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 94234 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| 3C1 | |
| Mouse monoclonal antibody raised against a partial recombinant FOXQ1. | |
| FOXQ1 | |
| HFH1 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.