missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FRS2, Mouse, Clone: S1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FRS2.
Brand: Abnova H00010818-M02A.200uL
This item is not returnable.
View return policy
Description
Sequence: MGSCCSCPDKDTVPDNHRNKFKVINVDDDGNELGSGIMELTDTELILYTRKRDSVKWHYLCLRRYGYDSNLFSFESGRRCQTGQGIFAFKCARAEELFNMSQEIMQNNSINVVEEPVVERNNHQTELEVPRTPRTPTTPGFAAQNLPNGYPRYPSFGDASSHPSSRHPSVGSARLPSVGEESTHPLLVAEEQVHTYVNTTGVQEERKNRTSVHVPLEARVSNAESSTPKEEPSSIEDRDPQILLEPEGVKFVLGPTPVQKQLMEKEKLEQLGRDQVSGSGANNTEWDTGYDSDERRDAPSVNKLVYENINGLSIPSASGVRRGRLTSTSTSDTQNINNSAQSpecifications
| FRS2 | |
| Monoclonal | |
| Unconjugated | |
| ascites with no preservative | |
| BC021562.1 | |
| FRS2 | |
| FRS2 (AAH21562.1,1 a.a. ∽ 341 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 10818 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Ascites |
| ELISA, Western Blot | |
| S1 | |
| Mouse monoclonal antibody raised against a partial recombinant FRS2. | |
| FRS2 | |
| FRS2A/FRS2alpha/SNT/SNT-1/SNT1 | |
| Mouse | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction