Learn More
FSHB, Mouse, Clone: 090-17275, Abnova™
Mouse monoclonal antibody raised against FSHB.
Brand: Abnova MAB4075.1mg
Description
The pituitary glycoprotein hormone family includes follicle-stimulating hormone, luteinizing hormone, chorionic gonadotropin, and thyroid-stimulating hormone. All of these glycoproteins consist of an identical alpha subunit and a hormone-specific beta subunit. This gene encodes the beta subunit of follicle-stimulating hormone. In conjunction with luteinizing hormone, follicle-stimulating hormone induces egg and sperm production. Alternative splicing results in two transcript variants encoding the same protein. [provided by RefSeq
Specifications
| FSHB | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| FSHB | |
| Mouse | |
| 1 mg | |
| Primary | |
| Follicle Stimulating Hormone (FSH). | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
| ELISA, Sandwich ELISA | |
| 090-17275 | |
| Mouse monoclonal antibody raised against FSHB. | |
| In 10mM phosphate buffer, 150mM NaCl, pH 7.4 (0.09% sodium azide) | |
| FSHB | |
| Human Follicle Stimulating Hormone (FSH). | |
| RUO | |
| 2488 | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.