Learn More
FUBP1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant FUBP1.
Brand: Abnova H00008880-A01.50uL
Description
This gene encodes a ssDNA binding protein that activates the far upstream element (FUSE) of c-myc and stimulates expression of c-myc in undifferentiated cells. Regulation of FUSE by FUBP occurs through single-strand binding of FUBP to the noncoding strand. This protein has been shown to function as an ATP-dependent DNA helicase. [provided by RefSeq
Sequence: VNDAFKDALQRARQIAAKIGGDAGTSLNSNDYGYGGQKRPLEDGDQPDAKKVAPQNDSFGTQLPPMHQQQSRSVMTEEYKVPDGMVGFIIGRGGEQISRIQQESGCKIQISpecifications
| FUBP1 | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a partial recombinant FUBP1. | |
| FUBP1 | |
| FBP/FUBP | |
| Mouse | |
| 50 μL | |
| Primary | |
| Human | |
| Antisera |
| ELISA, Western Blot | |
| Unconjugated | |
| 50% glycerol | |
| NM_003902 | |
| FUBP1 | |
| FUBP1 (NP_003893, 27 a.a. to 136 a.a) partial recombinant protein with GST tag. | |
| RUO | |
| 8880 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only