missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FUT4, Mouse, Clone: MMA or Leu-M1, Abnova™
Mouse monoclonal antibody raised against FUT4.
Brand: Abnova MAB11295.100ug
This item is not returnable.
View return policy
Description
The product of this gene transfers fucose to N-acetyllactosamine polysaccharides to generate fucosylated carbohydrate structures. It catalyzes the synthesis of the non-sialylated antigen, Lewis x (CD15). [provided by RefSeq
Specifications
| FUT4 | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (0.5-1ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| FUT4 | |
| FUT4 | |
| The U937 histiocytic cell line. | |
| RUO | |
| 2526 | |
| Store at 4°C. | |
| IgM κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
| MMA or Leu-M1 | |
| Mouse monoclonal antibody raised against FUT4. | |
| Bioreactor concentrate (0.05% sodium azide) | |
| CD15/ELFT/FCT3A/FUC-TIV/FUTIV | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction